Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atad5b
Entrez Gene ID
321619
Gene Name
ATPase family AAA domain containing 5b
Gene Aliases
dkeyp-97g3.4, fb23b03, rnf135, wu:fb23b03, zgc:66453
Chromosome Location
Chr.6: 76539 - 86646 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082829.2 NP_001076298.2 12 - 13 3084 62
XM_005166177.4 XP_005166234.1 12 - 13 3140 62
GenBank mRNA BC057535.1 - 12 - 13 3084 62

View Full Product Details