Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cisd2
Entrez Gene ID
393354
Gene Name
CDGSH iron sulfur domain 2
Gene Aliases
zgc:64148
Chromosome Location
Chr.1: 43686287 - 43692141 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200383.1 NP_956677.1 1 - 2 192 88
GenBank mRNA AY394933.1 - 1 - 2 195 88
BC053280.1 - 1 - 2 192 88

View Full Product Details