Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tfap2e
Entrez Gene ID
393794
Gene Name
transcription factor AP-2 epsilon
Gene Aliases
zgc:65882, zgc:77338
Chromosome Location
Chr.19: 47430954 - 47456898 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200821.2 NP_957115.2 5 - 6 974 83
XM_021468018.1 XP_021323693.1 5 - 6 1012 83
GenBank mRNA BC059800.1 - 5 - 6 942 83
EH595894.1 - 2 - 3 715 83

View Full Product Details