Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cap2
Entrez Gene ID
393809
Gene Name
cyclase associated actin cytoskeleton regulatory protein 2
Gene Aliases
zgc:113089, zgc:73317
Chromosome Location
Chr.19: 46158080 - 46210914 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200836.2 NP_957130.2 12 - 13 1504 80
XM_017352206.2 XP_017207695.1 12 - 13 1489 80
GenBank mRNA BC060935.1 - 12 - 13 1414 80
BC091655.1 - 12 - 13 1509 80

View Full Product Details