Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp2a2a
Entrez Gene ID
393940
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2a
Gene Aliases
atp2a2, zgc:55380
Chromosome Location
Chr.8: 11687231 - 11744993 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200965.1 NP_957259.1 20 - 21 3072 78
XM_005166798.4 XP_005166855.1 20 - 21 3098 78
XM_005166799.4 XP_005166856.1 20 - 21 3094 78
XM_021478220.1 XP_021333895.1 20 - 21 3098 78
GenBank mRNA BC045327.1 - 20 - 21 3072 78
BC165245.1 - 19 - 20 2891 78

View Full Product Details