Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
yy1a
Entrez Gene ID
322397
Gene Name
YY1 transcription factor a
Gene Aliases
fb59g10, wu:fa16g07, wu:fb59g10, yy1
Chromosome Location
Chr.17: 30857011 - 30863252 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212617.2 NP_997782.1 4 - 5 1014 77
GenBank mRNA AF389345.1 - - 1885 77
BC071351.1 - 4 - 5 1014 77

View Full Product Details