Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gnmt
Entrez Gene ID
403338
Gene Name
glycine N-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.17: 48921829 - 48935872 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212816.1 NP_997981.1 2 - 3 412 94
GenBank mRNA BC062527.1 - 2 - 3 412 94

View Full Product Details