Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
usf1l
Entrez Gene ID
406475
Gene Name
upstream transcription factor 1, like
Gene Aliases
usf2, wu:fb22c09, zgc:77845
Chromosome Location
Chr.5: 155327 - 167105 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213194.1 NP_998359.1 6 - 7 544 85
XM_005171828.4 XP_005171885.1 6 - 7 587 85
GenBank mRNA BC065876.1 - 6 - 7 544 85

View Full Product Details