Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nudt5
Entrez Gene ID
415176
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 5
Gene Aliases
wu:fe48a12, zgc:86930
Chromosome Location
Chr.4: 7870384 - 7876005 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002086.1 NP_001002086.1 4 - 5 316 73
GenBank mRNA BC071544.1 - 4 - 5 316 73

View Full Product Details