Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ddt
Entrez Gene ID
415237
Gene Name
D-dopachrome tautomerase
Gene Aliases
wu:fb49f11, zgc:86714
Chromosome Location
Chr.21: 15709112 - 15711760 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002147.1 NP_001002147.1 1 - 2 143 106
GenBank mRNA BC071391.1 - 1 - 2 143 106

View Full Product Details