Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrps23
Entrez Gene ID
445221
Gene Name
mitochondrial ribosomal protein S23
Gene Aliases
wu:fa18c12, zgc:100967
Chromosome Location
Chr.15: 30350054 - 30354786 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003615.1 NP_001003615.1 4 - 5 541 107
GenBank mRNA BC077090.1 - 4 - 5 541 107

View Full Product Details