Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbp2
Entrez Gene ID
445505
Gene Name
fructose-1,6-bisphosphatase 2
Gene Aliases
zgc:101083
Chromosome Location
Chr.5: 44813665 - 44829728 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004008.1 NP_001004008.1 1 - 2 220 72
XM_005165300.4 XP_005165357.1 1 - 2 326 72
GenBank mRNA BC080232.1 - 1 - 2 220 72

View Full Product Details