Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fgf14
Entrez Gene ID
497642
Gene Name
fibroblast growth factor 14
Gene Aliases
-
Chromosome Location
Chr.9: 31835221 - 32051533 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012382.1 NP_001012382.1 3 - 4 411 128
XM_005167695.4 XP_005167752.1 3 - 4 238 128
XM_009304735.3 XP_009303010.1 3 - 4 1224 128
XM_021478646.1 XP_021334321.1 3 - 4 361 128
XM_021478647.1 XP_021334322.1 3 - 4 239 128
GenBank mRNA AB195620.1 - 3 - 4 411 128
BC162852.1 - 3 - 4 477 128
BC162900.1 - 3 - 4 477 128

View Full Product Details