Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gpx7
Entrez Gene ID
552981
Gene Name
glutathione peroxidase 7
Gene Aliases
im:6902827, zgc:112293
Chromosome Location
Chr.23: 42219243 - 42231740 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020501.1 NP_001018337.1 1 - 2 159 74
GenBank mRNA BC095737.1 - 1 - 2 159 74

View Full Product Details