Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dcdc2b
Entrez Gene ID
563259
Gene Name
doublecortin domain containing 2B
Gene Aliases
zgc:123267
Chromosome Location
Chr.13: 33301899 - 33314758 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037689.2 NP_001032778.2 9 - 10 895 72
XM_017358759.2 XP_017214248.1 11 - 12 1282 72
XM_017358760.2 XP_017214249.1 11 - 12 1256 72
XM_017358761.2 XP_017214250.1 11 - 12 1269 72
XM_017358762.2 XP_017214251.1 11 - 12 1243 72
XM_017358763.2 XP_017214252.1 11 - 12 1261 72
XM_017358764.2 XP_017214253.1 11 - 12 1222 72
XM_017358765.2 XP_017214254.1 11 - 12 1282 72
XM_021480609.1 XP_021336284.1 10 - 11 2656 72
GenBank mRNA BC108062.1 - 9 - 10 906 72

View Full Product Details