Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plag1
Entrez Gene ID
561450
Gene Name
pleiomorphic adenoma gene 1
Gene Aliases
-
Chromosome Location
Chr.7: 58730201 - 58741500 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039814.1 NP_001034903.1 2 - 3 475 68
GenBank mRNA AY864859.1 - 2 - 3 475 68
BC124082.1 - 2 - 3 415 68
BC164032.1 - 1 - 2 265 68

View Full Product Details