Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp3c4
Entrez Gene ID
558541
Gene Name
cytochrome P450, family 3, subfamily C, polypeptide 4
Gene Aliases
zgc:153269
Chromosome Location
Chr.3: 40936094 - 40945724 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077548.1 NP_001071016.1 7 - 8 744 124
GenBank mRNA BC124295.1 - 7 - 8 744 124

View Full Product Details