Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk4
Entrez Gene ID
777730
Gene Name
cyclin-dependent kinase 4
Gene Aliases
zgc:153726
Chromosome Location
Chr.11: 135249 - 141592 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077777.1 NP_001071245.1 4 - 5 618 67
GenBank mRNA BC124438.1 - 4 - 5 618 67
BC164244.1 - 3 - 4 561 67

View Full Product Details