Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mxra8b
Entrez Gene ID
556385
Gene Name
matrix-remodelling associated 8b
Gene Aliases
zgc:153991
Chromosome Location
Chr.23: 29889160 - 29906158 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079960.3 NP_001073429.3 9 - 10 1385 90
GenBank mRNA BC124787.1 - 9 - 10 1354 90

View Full Product Details