Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lcor
Entrez Gene ID
558560
Gene Name
ligand dependent nuclear receptor corepressor
Gene Aliases
fd05a07, wu:fd05a07, zgc:158268
Chromosome Location
Chr.22: 34784070 - 34824240 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021469635.1 XP_021325310.1 5 - 6 693 98
XM_021469636.1 XP_021325311.1 4 - 5 616 98
GenBank mRNA BC127573.1 - 5 - 6 760 98

View Full Product Details