Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sigirr
Entrez Gene ID
563606
Gene Name
single immunoglobulin and toll-interleukin 1 receptor (TIR) domain
Gene Aliases
sc:d148
Chromosome Location
Chr.25: 22107606 - 22146307 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098250.1 NP_001091720.1 7 - 8 1024 122
GenBank mRNA BC129175.1 - 7 - 8 1025 122
CU458918.1 - 7 - 8 1024 122
JF714479.1 - 7 - 8 929 122
KJ574205.1 - 7 - 8 1013 122

View Full Product Details