Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
vsig10
Entrez Gene ID
100005564
Gene Name
V-set and immunoglobulin domain containing 10
Gene Aliases
fj20c09, si:ch73-13b6.3, wu:fj20c09
Chromosome Location
Chr.5: 2187 - 26236 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100094.2 NP_001093564.2 5 - 6 1466 118
GenBank mRNA CU638770.1 - 5 - 6 1466 118

View Full Product Details