Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adkb
Entrez Gene ID
322229
Gene Name
adenosine kinase b
Gene Aliases
cb980, fb53h03, wu:fb53h03
Chromosome Location
Chr.12: 46842395 - 46868003 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198802.1 NP_942097.1 9 - 10 922 109
GenBank mRNA BC051621.1 - 9 - 10 922 109

View Full Product Details