Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slc15a4
Entrez Gene ID
393932
Gene Name
solute carrier family 15 member 4
Gene Aliases
bZ1L10.1, zgc:56484, zgc:63767
Chromosome Location
Chr.8: 35809055 - 35961008 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320529.1 NP_001307458.1 6 - 7 1495 70
GenBank mRNA BC049320.1 - 8 - 9 1356 70
BC056821.1 - 7 - 8 1287 70

View Full Product Details