Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pigf
Entrez Gene ID
402942
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class F
Gene Aliases
zgc:77184
Chromosome Location
Chr.12: 25097806 - 25110974 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205645.1 NP_991208.1 4 - 5 632 73
GenBank mRNA BC065590.1 - 4 - 5 632 73

View Full Product Details