Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
otud3
Entrez Gene ID
405858
Gene Name
OTU deubiquitinase 3
Gene Aliases
zgc:85911
Chromosome Location
Chr.23: 39611750 - 39626681 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_212922.1 NP_998087.1 3 - 4 488 75
GenBank mRNA BC067689.1 - 3 - 4 488 75

View Full Product Details