Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxw11b
Entrez Gene ID
406825
Gene Name
F-box and WD repeat domain containing 11b
Gene Aliases
btrc2, fbxw11a, wu:fa12e12, wu:fb11f03, zgc:63728
Chromosome Location
Chr.10: 307452 - 333377 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213504.2 NP_998669.2 4 - 5 553 62
XM_005155294.4 XP_005155351.1 4 - 5 564 62
GenBank mRNA BC056809.1 - 4 - 5 555 62

View Full Product Details