The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098188.1 | NP_001091658.1 | 1 - 2 | 112 | 84 |
GenBank mRNA | AB175620.1 | - | 1 - 2 | 102 | 84 |
BC139601.1 | - | 1 - 2 | 112 | 84 | |
BC162603.1 | - | 1 - 2 | 102 | 84 | |
BC162606.1 | - | 1 - 2 | 102 | 84 |