Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fabp6
Entrez Gene ID
415166
Gene Name
fatty acid binding protein 6, ileal (gastrotropin)
Gene Aliases
zgc:92421
Chromosome Location
Chr.21: 30043037 - 30047356 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002076.2 NP_001002076.1 3 - 4 388 97
GenBank mRNA BC072553.1 - 3 - 4 371 97
EH493621.1 - 3 - 4 421 97
EH505333.1 - 3 - 4 417 97

View Full Product Details