Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
luc7l
Entrez Gene ID
415206
Gene Name
LUC7-like (S. cerevisiae)
Gene Aliases
wu:fb51e07, wu:fz47d12, zgc:101760, zgc:86815
Chromosome Location
Chr.24: 39638546 - 39654132 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002116.1 NP_001002116.1 9 - 10 1070 62
XM_005162898.3 XP_005162955.2 10 - 11 1183 62
XM_009297653.1 XP_009295928.1 9 - 10 1487 62
XM_009297654.2 XP_009295929.1 11 - 12 1483 62
XM_021470132.1 XP_021325807.1 11 - 12 1493 62
XM_021470133.1 XP_021325808.1 12 - 13 1393 62
XM_021470134.1 XP_021325809.1 11 - 12 1264 62
XM_021470135.1 XP_021325810.1 9 - 10 1268 62
GenBank mRNA BC071474.1 - 9 - 10 1070 62
BC083270.1 - 8 - 9 1231 62

View Full Product Details