Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
slc17a9b
Entrez Gene ID
436908
Gene Name
solute carrier family 17 member 9b
Gene Aliases
sb:cb849, wu:fa99h02, zgc:92222
Chromosome Location
Chr.23: 17522949 - 17549145 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002635.1 NP_001002635.1 3 - 4 705 64
GenBank mRNA BC075944.1 - 3 - 4 705 64

View Full Product Details