Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
echs1
Entrez Gene ID
368912
Gene Name
enoyl CoA hydratase, short chain, 1, mitochondrial
Gene Aliases
fj55e05, si:zc217g15.1, wu:fj55e05
Chromosome Location
Chr.13: 24579108 - 24582236 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004529.2 NP_001004529.1 5 - 6 623 101

View Full Product Details