Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
synpr
Entrez Gene ID
368919
Gene Name
synaptoporin
Gene Aliases
si:ch211-263a23.1, si:zc263a23.1, wu:fq14d10
Chromosome Location
Chr.6: 41223204 - 41229787 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004532.2 NP_001004532.1 3 - 4 425 124
GenBank mRNA BC086853.1 - 3 - 4 425 124
BC164641.1 - 3 - 4 372 124

View Full Product Details