Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
alkbh6
Entrez Gene ID
317736
Gene Name
alkB homolog 6
Gene Aliases
zgc:86795
Chromosome Location
Chr.5: 36650096 - 36654698 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005390.1 NP_001005390.1 1 - 2 241 120
GenBank mRNA BC071457.1 - 1 - 2 241 120

View Full Product Details