Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ankrd45
Entrez Gene ID
553808
Gene Name
ankyrin repeat domain 45
Gene Aliases
si:dkey-4i23.6, zgc:112418
Chromosome Location
Chr.2: 35728033 - 35731776 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020770.1 NP_001018606.1 2 - 3 458 86
GenBank mRNA BC095811.1 - 2 - 3 458 86

View Full Product Details