Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ankrd45
Entrez Gene ID
553808
Gene Name
ankyrin repeat domain 45
Gene Aliases
si:dkey-4i23.6, zgc:112418
Chromosome Location
Chr.2: 35728033 - 35731776 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020770.1 NP_001018606.1 4 - 5 688 103
GenBank mRNA BC095811.1 - 4 - 5 688 103

View Full Product Details