Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gtpbp1l
Entrez Gene ID
402863
Gene Name
GTP binding protein 1, like
Gene Aliases
-
Chromosome Location
Chr.22: 14836204 - 14851699 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024811.1 NP_001019982.1 6 - 7 1084 119
XM_005161694.4 XP_005161751.1 7 - 8 1139 119
XM_009296201.3 XP_009294476.1 7 - 8 1099 119
GenBank mRNA BC056806.1 - 6 - 7 1082 119
BC096803.1 - 6 - 7 1084 119

View Full Product Details