The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024811.1 | NP_001019982.1 | 9 - 10 | 1719 | 64 |
XM_005161694.4 | XP_005161751.1 | 10 - 11 | 1774 | 64 | |
XM_009296201.3 | XP_009294476.1 | 10 - 11 | 1734 | 64 | |
GenBank mRNA | BC056806.1 | - | 9 - 10 | 1717 | 64 |
BC096803.1 | - | 9 - 10 | 1719 | 64 |