Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
herpud1
Entrez Gene ID
565499
Gene Name
homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-like domain member 1
Gene Aliases
zgc:114134
Chromosome Location
Chr.18: 17600644 - 17608578 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025537.1 NP_001020708.1 3 - 4 441 111
GenBank mRNA BC097190.1 - 3 - 4 441 111

View Full Product Details