Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
inppl1a
Entrez Gene ID
325179
Gene Name
inositol polyphosphate phosphatase-like 1a
Gene Aliases
INPPL1-A, SHIP-2A, fc58a04, ship2a, wu:fc58a04, zgc:198309
Chromosome Location
Chr.15: 47454443 - 47498275 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039804.2 NP_001034893.1 13 - 14 2174 115
GenBank mRNA BC171582.1 - 14 - 15 2094 115
DQ272661.2 - 13 - 14 2174 115

View Full Product Details