Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mif
Entrez Gene ID
751093
Gene Name
macrophage migration inhibitory factor
Gene Aliases
-
Chromosome Location
Chr.14: 48964678 - 48970210 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043321.1 NP_001036786.1 2 - 3 405 105
GenBank mRNA AB298722.1 - 2 - 3 302 105
BC049029.1 - 2 - 3 393 105
BC171420.1 - 2 - 3 393 105
BC171422.1 - 2 - 3 393 105
DQ639953.1 - 2 - 3 405 105

View Full Product Details