Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
usp49
Entrez Gene ID
559473
Gene Name
ubiquitin specific peptidase 49
Gene Aliases
si:rp71-11b17.7
Chromosome Location
Chr.22: 564638 - 581689 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044896.1 NP_001038361.1 4 - 5 1645 69
XM_005168272.4 XP_005168329.1 6 - 7 3350 69
XM_009295758.3 XP_009294033.1 6 - 7 3022 69
GenBank mRNA BC171546.1 - 4 - 5 1683 69
BC171548.1 - 4 - 5 1683 69

View Full Product Details