Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
usp49
Entrez Gene ID
559473
Gene Name
ubiquitin specific peptidase 49
Gene Aliases
si:rp71-11b17.7
Chromosome Location
Chr.22: 564638 - 581689 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044896.1 NP_001038361.1 5 - 6 1845 70
XM_005168272.4 XP_005168329.1 7 - 8 3550 70
XM_009295758.3 XP_009294033.1 7 - 8 3222 70
GenBank mRNA BC171546.1 - 5 - 6 1883 70
BC171548.1 - 5 - 6 1883 70

View Full Product Details