Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
skor1b
Entrez Gene ID
567457
Gene Name
SKI family transcriptional corepressor 1b
Gene Aliases
lbxcor1, si:dkey-246a16.3, skor1
Chromosome Location
Chr.18: 168179 - 173867 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045133.1 NP_001038598.1 1 - 2 1726 66
XM_005159134.4 XP_005159191.1 2 - 3 2251 66
XM_005159135.4 XP_005159192.1 2 - 3 2250 66
XM_005159136.4 XP_005159193.1 2 - 3 2253 66
GenBank mRNA BC134846.1 - 2 - 3 2028 66

View Full Product Details