Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pstpip1a
Entrez Gene ID
571331
Gene Name
proline-serine-threonine phosphatase interacting protein 1a
Gene Aliases
si:dkey-24l11.5
Chromosome Location
Chr.18: 109909 - 137807 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045216.1 NP_001038681.1 2 - 3 132 73

View Full Product Details