Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pbrm1l
Entrez Gene ID
445141
Gene Name
polybromo 1, like
Gene Aliases
pb1, zgc:100857, zgc:113882
Chromosome Location
Chr.6: 251612 - 277433 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC098878.1 - 8 - 9 1337 145

View Full Product Details