Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
camsap1b
Entrez Gene ID
561094
Gene Name
calmodulin regulated spectrin-associated protein 1b
Gene Aliases
camsap1, si:dkey-157g20.3
Chromosome Location
Chr.5: 65613292 - 65663047 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100001.3 NP_001093471.1 9 - 10 1193 73
XM_005168648.4 XP_005168705.1 8 - 9 1211 73
XM_021476204.1 XP_021331879.1 9 - 10 1244 73

View Full Product Details