Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppm1ba
Entrez Gene ID
30672
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1Ba
Gene Aliases
pp2cb, zgc:92031
Chromosome Location
Chr.13: 10683845 - 10697156 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131398.1 NP_571473.1 5 - 6 1182 93
GenBank mRNA BC079530.1 - 5 - 6 1182 93

View Full Product Details