Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pcmt
Entrez Gene ID
30751
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase
Gene Aliases
etID309741.20, fj13d06, pimt, wu:fj13d06
Chromosome Location
Chr.20: 1293985 - 1314555 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131465.3 NP_571540.1 2 - 3 244 109
XM_005160307.4 XP_005160364.1 2 - 3 372 109
GenBank mRNA BC049067.1 - 2 - 3 240 109
BC075735.1 - 2 - 3 212 109
U37434.1 - 2 - 3 203 109

View Full Product Details