Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
scn8aa
Entrez Gene ID
58152
Gene Name
sodium channel, voltage gated, type VIII, alpha subunit a
Gene Aliases
Nav1.6, scn8a
Chromosome Location
Chr.23: 27248689 - 27345450 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_131628.1 NP_571703.1 16 - 17 3089 80
XM_005162268.4 XP_005162325.1 16 - 17 3133 80
XM_009296871.3 XP_009295146.1 16 - 17 3133 80
XM_009296874.3 XP_009295149.1 16 - 17 3099 80
XM_021469841.1 XP_021325516.1 16 - 17 3278 80
XM_021469842.1 XP_021325517.1 16 - 17 3147 80
XM_021469843.1 XP_021325518.1 16 - 17 3146 80
GenBank mRNA AF297658.1 - 16 - 17 3089 80
BC163773.1 - 16 - 17 2943 80

View Full Product Details